Antimony, Oil based standard solution, Specpure®, Sb 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042433-18
AA42433-18EA
66.2
USD
AA42433-18
Antimony, Oil based standard solution, Specpure®, Sb 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Antimony Standards
Physical Form: Liquid
MDL No.: MFCD00134030
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...