Chlorodimethyloctadecylsilane ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
042414-14
042414-22
042414-36
042414-06
AA42414-14EA
85.6
USD
AA42414-14
AA42414-22
AA42414-36
AA42414-06
Chlorodimethyloctadecylsilane ≥95%
Chlorodimethyloctadecylsilane
Preparation of hydrophobic surfaces, chain terminator
Purity: 95%
CAS No.: 18643-08-8
Molecular Formula: C20H43ClSi
Formula Weight: 347.10
Flash Point: 200°(392°F)
Physical Form: Solid
Density: 0.850
Refractive Index: 1.4530
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Acrid
Melting Point: 28-32°
Boiling Point: 184-186°/0.2mm
Beilstein Reference: 2246012
UN No.: 3261
MDL No.: MFCD00000504
Preparation of hydrophobic surfaces, chain terminator
Formula:
C₂₀H₄₃ClSi MW: 347.10 g/mol Boiling Pt: 184…186 °C (0.2 mmHg) Melting Pt: 28…32 °C Density: 0.850 Flash Pt: 200 °C (392 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000504 CAS Number: 18643-08-8 EINECS: 242-472-2 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...