Deuterium chloride 20% (w/w) in D₂O (99.5% D) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

042407-18
AA42407-18EA 181 USD
AA42407-18
Deuterium chloride 20% (w/w) in D₂O (99.5% D) (w/w)
Deuterium chloride

MDL: MFCD00044642


Purity: 99.5% CAS No.: 7698-05-7 Molecular Formula: DCl Formula Weight: 37.47 Physical Form: Liquid, in clear glass ampoules Density: 1.20 Refractive Index: 1.3730 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Odor: Acidic Boiling Point: 108° UN No.: 1789 MDL No.: MFCD00044642

Formula: DCl
Boiling Pt: 108 °C
Density: 1.20
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR