Deuterium chloride 20% (w/w) in D₂O (99.96% D)
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00044642
Purity: 99.96+%
CAS No.: 7698-05-7
Molecular Formula: DCl
Formula Weight: 37.47
Physical Form: Liquid, in clear glass ampoules
Density: 1.20
Refractive Index: 1.3730
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
Odor: Acidic
Boiling Point: 108°
UN No.: 1789
MDL No.: MFCD00044642
Formula:
DCl Boiling Pt: 108 °C Density: 1.20 Storage Temperature: Ambient |
MDL Number:
MFCD00044642 CAS Number: 7698-05-7 EINECS: 231-715-8 UN: 1789 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...