Cobalt ≥99.95% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals
Danger

042354-14 042354-22 042354-06
AA42354-14EA 373 USD
AA42354-14 AA42354-22 AA42354-06
Cobalt ≥99.95% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 3.175 mm (0.125 in)
Cobalt

MDL: MFCD00010935


Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈0.45g/slug Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR