Tetrahydrofuran-[D₈] (100% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8
042348-KV
AA42348-KVEA
836
USD
AA42348-KV
Tetrahydrofuran-[D₈] (100% D) (Isotopic) for NMR spectroscopy
Tetrahydrofuran-[D8]
MDL: MFCD00044238
Purity: 100%
CAS No.: 1693-74-9
Molecular Formula: C4D8O
Formula Weight: 80.16
Flash Point: -17°(-1°F)
Physical Form: Liquid in prescored ampoules, 0.75ml/ampoule
Density: 0.985
Appearance: Colorless
Melting Point: -106°
Boiling Point: 66°
UN No.: 2056
MDL No.: MFCD00044238
Formula:
C₄D₈O MW: 80.04 g/mol Boiling Pt: 66 °C Melting Pt: –106 °C Density: 0.985 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044238 CAS Number: 1693-74-9 EINECS: 216-898-4 UN: 2056 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...