Dichloromethane-[D₂] (100% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

042334-KV
AA42334-KVEA 725 USD
AA42334-KV AA42334-SA
Dichloromethane-[D₂] (100% D) (Isotopic) for NMR spectroscopy
Dichloromethane-[D2]

MDL: MFCD00000882 Notes: Minimum isotopic purity 99.96%


Purity: 100% CAS No.: 1665-00-5 Molecular Formula: CD2Cl2 Formula Weight: 86.95 Physical Form: Liquid in prescored ampoules, 0.75ml/ampoule Density: 1.35 UN No.: 1593 MDL No.: MFCD00000882

Formula: CD₂Cl₂
Boiling Pt: 40 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.35
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR