N,N-Dimethylformamide-[D₇] (99.5% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7

042284-03 042284-46 042284-06 042284-09
AA42284-03EA 80.89 USD
AA42284-03 AA42284-46 AA42284-06 AA42284-09
N,N-Dimethylformamide-[D₇] (99.5% D) (Isotopic) for NMR spectroscopy
N,N-Dimethylformamide-[D7]

MDL: MFCD00003286


Purity: 99.5% CAS No.: 4472-41-7 Molecular Formula: C3D7NO Formula Weight: 80.14 Flash Point: 58°(136°F) Physical Form: Liquid in prescored ampoules Density: 1.04 Appearance: Colorless Odor: Amine-like Boiling Point: 153° UN No.: 2265 MDL No.: MFCD00003286

Formula: DCOON(CD₃)₂
Boiling Pt: 153 °C
Melting Pt: –60 °C
Density: 1.04
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR