Acetone-[D₆] (100% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6
042263-SA
AA42263-SAEA
77.6
USD
AA42263-SA
Acetone-[D₆] (100% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Acetone-[D6]
MDL: MFCD00044635
Notes: Minimum isotopic purity 99.96%
Purity: 100%
CAS No.: 666-52-4
Molecular Formula: CD3COCD3
Formula Weight: 64.13
Flash Point: -17°(1°F)
Physical Form: Liquid in prescored ampoules, 0.75ml/ampoule
Density: 0.872
Refractive Index: 1.3554
Appearance: Colorless
Odor: Acetone-like
Melting Point: -94°
Boiling Point: 56°
UN No.: 1090
MDL No.: MFCD00044635
Formula:
D₃CC(O)CD₃ MW: 64.03 g/mol Boiling Pt: 56 °C Melting Pt: –94 °C Density: 0.872 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044635 CAS Number: 666-52-4 EINECS: 211-563-9 UN: 1090 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...