Osmium, AAS standard solution, Specpure®, Os 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042239-AE
AA42239-APEA
416.25
USD
AA42239-AP
AA42239-AE
Osmium, AAS standard solution, Specpure®, Os 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Osmium Standards
Molecular Formula: (NH4)2OsCl6 in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011147
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...