Potassium fluoride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
042217-14
AA42217-14EA
222
USD
AA42217-14
Potassium fluoride ≥99.9% (metals basis)
Potassium fluoride
MDL: MFCD00011398
Beilstein Registry No.: 3902818
Purity: 99.9%
CAS No.: 7789-23-3
Molecular Formula: FK
Formula Weight: 58.10
Physical Form: Crystalline
Density: 2.48
Refractive Index: 1.363
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 859°
Boiling Point: 1505°
Beilstein Reference: 3902818
Merck Reference: 14,7632
UN No.: 1812
MDL No.: MFCD00011398
Formula:
KF MW: 58.1 g/mol Boiling Pt: 1505 °C Melting Pt: 859 °C Density: 2.48 Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 14,7632 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...