Lauric acid ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00002736
Beilstein Registry No.: 1099477
Purity: 99.5%
CAS No.: 143-07-7
Molecular Formula: CH3(CH2)10CO2H
Formula Weight: 200.32
Flash Point: 165°(329°F)
Physical Form: Crystalline
Density: 0.883
Odor: Odorless
Melting Point: 43-45°
Boiling Point: 180°/21mm
Beilstein Reference: 1099477
Merck Reference: 14,5384
MDL No.: MFCD00002736
Formula:
C₁₂H₂₄O₂ Boiling Pt: 180 °C (21 mmHg) Melting Pt: 43…45 °C Density: 0.883 Flash Pt: 165 °C (329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002736 CAS Number: 143-07-7 EINECS: 205-582-1 Merck Index: 14,5384 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...