Magnesium acrylate

Supplier: Thermo Scientific Chemicals
Warning

042002-14 042002-06 042002-22
AA42002-14EA 120 USD
AA42002-14 AA42002-06 AA42002-22
Magnesium acrylate
Magnesium acrylate

CAS No.: 5698-98-6 Molecular Formula: C6H6MgO4 Formula Weight: 166.42 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Melting Point: 220° dec. MDL No.: MFCD00068285

Formula: C₆H₆MgO₄
Melting Pt: >220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00068285
CAS Number: 5698-98-6
EINECS: 227-177-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR