Magnesium acrylate
Supplier: Thermo Scientific Chemicals
|
Warning
|
042002-14
042002-06
042002-22
AA42002-14EA
120
USD
AA42002-14
AA42002-06
AA42002-22
Magnesium acrylate
Magnesium acrylate
CAS No.: 5698-98-6
Molecular Formula: C6H6MgO4
Formula Weight: 166.42
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Melting Point:
Formula:
C₆H₆MgO₄ Melting Pt: >220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00068285 CAS Number: 5698-98-6 EINECS: 227-177-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...