Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hafnium(IV) t-butoxide, Hafnium(IV)-tert-butanolate

041908-04 041908-09 041908-18
AA41908-04EA 131 USD
AA41908-04 AA41908-09 AA41908-18
Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Hafnium(IV) tert-butoxide

Purity: 99.9% CAS No.: 2172-02-3 Molecular Formula: C16H36HfO4 Formula Weight: 470.94 Flash Point: 28°(82°F) Physical Form: Liquid Density: 1.15 Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow Melting Point: 2° Boiling Point: 90°/5mm Solubility: Soluble in hydrocarbons, reacts with alcohols, ketones, and esters UN No.: 1993 MDL No.: MFCD00070458

Formula: C₁₆H₃₆HfO₄
Boiling Pt: 90 °C (5 mmHg)
Melting Pt: 2 °C
Density: 1.15
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070458
CAS Number: 2172-02-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR