Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hafnium(IV) t-butoxide, Hafnium(IV)-tert-butanolate
041908-04
041908-09
041908-18
AA41908-04EA
131
USD
AA41908-04
AA41908-09
AA41908-18
Hafnium(IV) tert-butoxide ≥99.9% (metals basis excluding Zr) (max. 0.5% Zr)
Hafnium(IV) tert-butoxide
Purity: 99.9%
CAS No.: 2172-02-3
Molecular Formula: C16H36HfO4
Formula Weight: 470.94
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 1.15
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow
Melting Point: 2°
Boiling Point: 90°/5mm
Solubility: Soluble in hydrocarbons, reacts with alcohols, ketones, and esters
UN No.: 1993
MDL No.: MFCD00070458
Formula:
C₁₆H₃₆HfO₄ Boiling Pt: 90 °C (5 mmHg) Melting Pt: 2 °C Density: 1.15 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070458 CAS Number: 2172-02-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...