Polyimide, film, Thickness 0.013 mm

Supplier: Thermo Scientific Chemicals

041897-GH 041897-GT
AA41897-GHEA 262 USD
AA41897-GH AA41897-GT
Polyimide, film, Thickness 0.013 mm
Polyimide

CAS No.: 62929-02-6 Physical Form: ≈0.1846g/100x100mm MDL No.: MFCD00677744

Formula: C₃₅H₂₈N₂O₇
Storage Temperature: Ambient
MDL Number: MFCD01868102
CAS Number: 62929-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR