Cetrimonium chloride ≥96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC

041861-06 041861-14 041861-22 041861-36
AA41861-06EA 38.8 USD
AA41861-06 AA41861-14 AA41861-22 AA41861-36
Cetrimonium chloride ≥96%
Cetrimonium chloride

MDL: MFCD00011773 Beilstein Registry No.: 3657974


Purity: 96% CAS No.: 112-02-7 Molecular Formula: C19H42ClN Formula Weight: 320.00 Physical Form: Powder Density: 0.968 Appearance: Off-white Odor: Odorless Melting Point: 234° Beilstein Reference: 3657974 UN No.: 1759 MDL No.: MFCD00011773

Formula: C₁₉H₄₂ClN
Melting Pt: 234 °C
Density: 0,968
Storage Temperature: Ambient
MDL Number: MFCD00011773
CAS Number: 112-02-7
EINECS: 203-928-6
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR