Cetrimonium chloride ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC
041861-06
041861-14
041861-22
041861-36
AA41861-06EA
38.8
USD
AA41861-06
AA41861-14
AA41861-22
AA41861-36
Cetrimonium chloride ≥96%
Cetrimonium chloride
MDL: MFCD00011773
Beilstein Registry No.: 3657974
Purity: 96%
CAS No.: 112-02-7
Molecular Formula: C19H42ClN
Formula Weight: 320.00
Physical Form: Powder
Density: 0.968
Appearance: Off-white
Odor: Odorless
Melting Point: 234°
Beilstein Reference: 3657974
UN No.: 1759
MDL No.: MFCD00011773
Formula:
C₁₉H₄₂ClN Melting Pt: 234 °C Density: 0,968 Storage Temperature: Ambient |
MDL Number:
MFCD00011773 CAS Number: 112-02-7 EINECS: 203-928-6 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...