tert-Butanol, anhydrous ≥99.8%, chemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
041857-M1
041857-K2
041857-AK
041857-AD
AA41857-M1EA
397
USD
AA41857-M1
AA41857-K2
AA41857-AK
AA41857-AD
tert-Butanol, anhydrous ≥99.8%, chemSeal™
tert-Butanol
MDL: MFCD00004464
Beilstein Registry No.: 906698
Notes: Water less than 0.01%
Formula:
(CH₃)₃COH Boiling Pt: 83 °C Melting Pt: 25 °C Density: 0.786 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,II Merck Index: 14,1542 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...