tert-Butanol, anhydrous ≥99.8%, chemSeal™

Supplier: Thermo Scientific Chemicals
Danger


Packaged under argon in resealable ChemSeal™ bottles.

041857-M1 041857-K2 041857-AK 041857-AD
AA41857-M1EA 397 USD
AA41857-M1 AA41857-K2 AA41857-AK AA41857-AD
tert-Butanol, anhydrous ≥99.8%, chemSeal™
tert-Butanol

MDL: MFCD00004464 Beilstein Registry No.: 906698 Notes: Water less than 0.01%

Formula: (CH₃)₃COH
Boiling Pt: 83 °C
Melting Pt: 25 °C
Density: 0.786
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
Merck Index: 14,1542
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR