tert-Butyl methyl ether, anhydrous 99+%, chemSeal™ for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
Chromatography solvent
Purity: 99+%
CAS No.: 1634-04-4
Molecular Formula: (CH3)3COCH3
Formula Weight: 88.15
Flash Point: -25°(-13°F)
Physical Form: Liquid
Density: 0.742
Refractive Index: 1.3690
Odor: Mild
Melting Point: -109°
Boiling Point: 55-56°
Beilstein Reference: 1730942
Merck Reference: 14,6032
UN No.: 2350
MDL No.: MFCD00008812
Chromatography solvent
Formula:
(CH₃)₃COCH₃ Boiling Pt: 55…56 °C Melting Pt: –109 °C Density: 0.742 Flash Pt: –25 °C (–13 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008812 CAS Number: 1634-04-4 EINECS: 216-653-1 UN: 2350 ADR: 3,II Merck Index: 14,6032 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...