Boron trifluoride diethyl ether complex (≥46,5% BF₃)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate

Packaged under Argon in resealable ChemSeal™ bottles.

041796-22 041796-36
AA41796-22EA 46.98 USD
AA41796-22 AA41796-36
Boron trifluoride diethyl ether complex (≥46,5% BF₃)
Boron trifluoride diethyl ether complex

CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Physical Form: Liquid Density: 1.130 Refractive Index: 1.3440 Sensitivity: Moisture Sensitive Odor: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Fieser: 1,70 13,43 14,54 15,45 16,44 17,52 18,60 19,46 20,55 21,66 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194 Packaged under Argon in resealable ChemSeal™ bottles

Formula: C₄H₁₀BF₃O
Boiling Pt: 125…126 °C
Melting Pt: –60 °C
Density: 1.130
Flash Pt: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013194
CAS Number: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8,I
Merck Index: 14,1350

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR