Silicon(II) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Silicium (II) oxide, Silicon monoxide, Silicon oxide

041710-22 041710-36
AA41710-22EA 211 USD
AA41710-22 AA41710-36
Silicon(II) oxide ≥99.9% (metals basis)
Silicon (II) oxide

For vacuum deposition


Purity: 99.9% CAS No.: 10097-28-6 Molecular Formula: SiO Formula Weight: 44.09 Physical Form: 3-6mm Pieces Density: 2.13 Melting Point: 1702° Boiling Point: 1880° Merck Reference: 14,8496 MDL No.: MFCD00151536


For vacuum deposition

Formula: SiO
MW: 44.08 g/mol
Boiling Pt: 1880
Melting Pt: 1702
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 14,8496

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR