Zirconium ≥99.5% (metals basis excluding Hf), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

041659-KS
AA41659-KSEA 297 USD
AA41659-KS
Zirconium ≥99.5% (metals basis excluding Hf), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Zirconium

Magnetron sputtering source


Purity: 99.5% CAS No.: 7440-67-7 Odor: Odorless MDL No.: MFCD00011303


Magnetron sputtering source

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR