Platinum ≥99.95% (metals basis), thin foil, hard, Thickness 0.00125 mm (0.00005 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

041632-FF
AA41632-FFEA 1879 USD
AA41632-FF
Platinum ≥99.95% (metals basis), thin foil, hard, Thickness 0.00125 mm (0.00005 in)
Platinum

MDL: MFCD00011179


Purity: 99.95% CAS No.: 7440-06-4 Molecular Formula: Not certified pinhole free Physical Form: ≈0.017g/25x25mm, Temper: as rolled Odor: Odorless

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR