Lanthanum ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

041558-FF
AA41558-FFEA 327 USD
AA41558-FF
Lanthanum ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.1 mm (0.004 in)
Lanthanum

MDL: MFCD00011066


Purity: 99.9% CAS No.: 7439-91-0 Physical Form: ≈0.39g/25x25mm, Packaged in mylar under argon Appearance: Gray to silver UN No.: 3208 MDL No.: MFCD00011066

Formula: La
Boiling Pt: 3464 °C (1013 hPa)
Melting Pt: 920 °C
Density: ∼6,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR