Bismuth ≥99.99% (metals basis), rods, Diameter 11 mm (0.43 in)
Supplier: Thermo Scientific Chemicals
041534-36
041534-14
041534-22
AA41534-36EA
858
USD
AA41534-36
AA41534-14
AA41534-22
Bismuth ≥99.99% (metals basis), rods, Diameter 11 mm (0.43 in)
Bismuth
MDL: MFCD00134033
Purity: 99.99%
CAS No.: 7440-69-9
Physical Form: ≈9.3g/cm
Odor: Odorless
MDL No.: MFCD00134033
Formula:
Bi Boiling Pt: 1490 °C (1013 hPa) Melting Pt: 271…272 °C Density: 9.78 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 Merck Index: 12,01297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...