Glassy carbon, spherical powder 630-1000 µm Type 2

Supplier: Thermo Scientific Chemicals
Warning

041530-30
AA41530-30EA 639 USD
AA41530-30
Glassy carbon, spherical powder 630-1000 µm Type 2
Glassy carbon

UN No.: 1325 MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1 III
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR