Bismuth ≥99.99% (metals basis), needles
Supplier: Thermo Scientific Chemicals
041485-22
041485-A3
041485-36
AA41485-22EA
62
USD
AA41485-22
AA41485-A3
AA41485-36
Bismuth ≥99.99% (metals basis), needles
Bismuth
MDL: MFCD00134033
Purity: 99.99%
CAS No.: 7440-69-9
Physical Form: Approx. 1.5-3.0mm diameter; Various lengths
Odor: Odorless
MDL No.: MFCD00134033
Formula:
Bi Boiling Pt: 1490 °C (1013 hPa) Melting Pt: 271…272 °C Density: 9.78 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 Merck Index: 12,01297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...