Manganese ≥99.6% (metals basis), powder -140+325 mesh

Supplier: Thermo Scientific Chemicals
Warning

041343-14 041343-22 041343-36
AA41343-14EA 38.2 USD
AA41343-14 AA41343-22 AA41343-36
Manganese ≥99.6% (metals basis), powder -140+325 mesh
Manganese

MDL: MFCD00011111


Purity: 99.6% CAS No.: 7439-96-5 UN No.: 3089 MDL No.: MFCD00011111

Formula: Mn
Boiling Pt: 2097 °C (1013 hPa)
Melting Pt: 1260 °C
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,05762

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR