4-(2-Hydroxyethyl)benzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
041283-03
041283-06
AA41283-03EA
216
USD
AA41283-03
AA41283-MD
AA41283-06
4-(2-Hydroxyethyl)benzonitrile 98%
4-(2-Hydroxyethyl)benzonitrile
Applications: Organic intermediate
Purity: 98%
CAS No.: 69395-13-7
Molecular Formula: C9H9NO
Formula Weight: 147.18
Physical Form: Powder/Chunk
Boiling Point: 116-118°/0.5mm
UN No.: 3439
MDL No.: MFCD00052461
Organic intermediate
Formula:
C₉H₉NO Boiling Pt: 116…118 °C (0.5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00052461 CAS Number: 69395-13-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...