Polyvinyl alcohol, M.W. ∼ 88,000-97,000 98-99%, high molecular weight, hydrolysed
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(vinyl alcohol), PVOH, PVA, Mowiol®
041243-22
041243-14
041243-36
AA41243-22EA
66.19
USD
AA41243-22
AA41243-14
AA41243-36
Polyvinyl alcohol, M.W. ∼ 88,000-97,000 98-99%, high molecular weight, hydrolysed
Polyvinyl alcohol
Average M.W. 88,000-97,000
Purity: 98-99%
CAS No.: 9002-89-5
Molecular Formula: [-CH2CH(OH)-]n
Physical Form: Solid
Odor: Odorless
MDL No.: MFCD00081922
Formula:
[-CH₂CH(OH)-]n Storage Temperature: Ambient |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...