Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

041097-KS
AA41097-KSEA 274 USD
AA41097-KS
Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 6.35 mm (0.250 in)
Silicium dioxide

Purity: 99.995% Molecular Formula: SiO2 Formula Weight: 60.09 MDL No.: MFCD00011232

Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1719 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR