Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

041096-KS
AA41096-KSEA 232 USD
AA41096-KS
Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Silicium dioxide

Applications: Magnetron sputtering source


Purity: 99.995% Molecular Formula: SiO2 Formula Weight: 60.09 MDL No.: MFCD00011232


Magnetron sputtering source

Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1719 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR