Magnesium oxide ≥99.95% (metals basis excluding Ca), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesia usta

041072-KS
AA41072-KSEA 436 USD
AA41072-KS
Magnesium oxide ≥99.95% (metals basis excluding Ca), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Magnesium oxide

Magnetron sputtering source


Purity: 99.95% Molecular Formula: MgO; Ca 750ppm max Formula Weight: 40.30 MDL No.: MFCD00011109


Magnetron sputtering source

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR