Glassy Carbon Electron Beam Evaporating Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
040965-KT
AA40960-KTEA 388 USD
AA40960-KT AA40961-KT AA40962-KT AA40965-KT AA40963-KT AA40964-KT
Glassy Carbon Electron Beam Evaporating Crucibles, Thermo Scientific
Crucibles
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
All vessels feature a 15° angle. These crucibles are designed for high-temperature differential thermal/thermal gravimetric analyses.
Alta Aesar's glassy carbon products are subjected to a high temperature heat treatment which imparts special material properties, including significantly improved corrosion resistance and strength.