Tungsten, gauze woven from wire 100 mesh, Diameter 0.0509 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

040925-FL 040925-GT 040925-HB
AA40925-FLEA 370 USD
AA40925-FL AA40925-GT AA40925-HB
Tungsten, gauze woven from wire 100 mesh, Diameter 0.0509 mm (0.002 in)
Tungsten

MDL: MFCD00011461 Notes: Open area: 64%; Width of opening: 0.008in


CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR