Tungsten, gauze woven from wire 100 mesh, Diameter 0.0254 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
040924-GT
040924-HB
040924-FL
AA40924-GTEA
571
USD
AA40924-GT
AA40924-HB
AA40924-FL
Tungsten, gauze woven from wire 100 mesh, Diameter 0.0254 mm (0.001 in)
Tungsten
MDL: MFCD00011461
Notes: Open area: 81%; Width of opening: 0.009in
CAS No.: 7440-33-7
Physical Form: Wire Cloth
MDL No.: MFCD00011461
Formula:
W Boiling Pt: 5660 °C (1013 hPa) Melting Pt: 3410 °C Density: 19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...