Tungsten, gauze woven from wire 30 mesh, Diameter 0.1018 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

040923-FL 040923-GT 040923-HB
AA40923-FLEA 175 USD
AA40923-FL AA40923-GT AA40923-HB
Tungsten, gauze woven from wire 30 mesh, Diameter 0.1018 mm (0.004 in)
Tungsten

MDL: MFCD00011461 Notes: Open area: 77.4%; Width of opening: 0.029in


CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR