Tin ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

040898-KS
AA40898-KSEA 181 USD
AA40898-KS
Tin ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Tin

Magnetron sputtering source


Purity: 99.995% CAS No.: 7440-31-5 Odor: Odorless MDL No.: MFCD00133862


Magnetron sputtering source

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR