Manganese ≥99.95% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

040863-KS
AA40863-KSEA 1188 USD
AA40863-KS
Manganese ≥99.95% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 6.35 mm (0.250 in)
Manganese

MDL: MFCD00011111 Notes: Thickness tolerance: +/- 0.025in; Diameter tolerance: +/- 0.020in


Purity: 99.95% CAS No.: 7439-96-5 Physical Form: Hot pressed Odor: Odorless MDL No.: MFCD00011111

Formula: Mn
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR