Magnesium ≥99.95% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

040861-KS
AA40861-KSEA 462 USD
AA40861-KS
Magnesium ≥99.95% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 6.35 mm (0.250 in)
Magnesium

MDL: MFCD00085308 Notes: Thickness tolerance: +/- 0.015in; Diameter tolerance: +0.020/-0.0in


Purity: 99.95% CAS No.: 7439-95-4 Odor: Odorless MDL No.: MFCD00085308

Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR