Chromium ≥99.95% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

040842-KS
AA40842-KSEA 403 USD
AA40842-KS
Chromium ≥99.95% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Chromium

Magnetron sputtering source


Purity: 99.95% CAS No.: 7440-47-3 Odor: Odorless MDL No.: MFCD00010944


Magnetron sputtering source

Formula: Cr
Boiling Pt: 2469 °C (1013 hPa)
Melting Pt: 1840 °C
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR