Carbon ≥99.999% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Charcoal

040830-KS
AA40830-KSEA 431 USD
AA40830-KS
Carbon ≥99.999% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)
Carbon

MDL: MFCD00133992 Notes: Thickness tolerance: +/- 0.015in; Diameter tolerance: +/- 0.020in


Purity: 99.999% Physical Form: Hot Pressed MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Boiling Pt: ∼4000 °C (1013 hPa)
Melting Pt: 3727
Density: 3.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-955-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR