Aluminium ≥99.99% (metals basis), foil, annealed, Thickness 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

040763-GH 040763-FI 040763-HP
AA40763-GHEA 214 USD
AA40763-GH AA40763-FI AA40763-HP
Aluminium ≥99.99% (metals basis), foil, annealed, Thickness 2.0 mm (0.08 in)
Aluminium

MDL: MFCD00134029


Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈13.5g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR