Ammonium iron (II) sulfate hexahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

040751-03 040751-06 040751-14
AA40751-03EA 23.54 USD
AA40751-03 AA40751-06 AA40751-14
Ammonium iron (II) sulfate hexahydrate ≥99.999% (metals basis), Puratronic®
Ammonium iron (II) sulphate hexahydrate

MDL: MFCD00150530


Purity: 99.999% CAS No.: 7783-85-9 Molecular Formula: H8FeN2O8S2.xH2O (x≈6) Formula Weight: 284.04(anhy) Physical Form: Coarse Powder Density: 1.864 Appearance: Pale bluish green Odor: Odorless Melting Point: ca 100° dec. Merck Reference: 14,521 MDL No.: MFCD00150530

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: ∼ 100 °C (dec.)
Density: 1.864
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-382-4
Merck Index: 14,521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR