Ammonium iron (II) sulfate hexahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate
MDL: MFCD00150530
Purity: 99.999%
CAS No.: 7783-85-9
Molecular Formula: H8FeN2O8S2.xH2O (x≈6)
Formula Weight: 284.04(anhy)
Physical Form: Coarse Powder
Density: 1.864
Appearance: Pale bluish green
Odor: Odorless
Melting Point: ca 100° dec.
Merck Reference: 14,521
MDL No.: MFCD00150530
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O Melting Pt: ∼ 100 °C (dec.) Density: 1.864 Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-382-4 Merck Index: 14,521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...