Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness 0.0075 micron

Supplier: Thermo Scientific Chemicals

040682-DH 040682-HF
AA40682-DHEA 915 USD
AA40682-DH AA40682-HF
Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness 0.0075 micron
Aluminium

Microelectronics, contacts and coatings


Purity: 99.995% CAS No.: 7429-90-5 Physical Form: Circular disc on permanent mylar support Odor: Odorless MDL No.: MFCD00134029


Microelectronics, contacts and coatings

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR