Niobium ≥99.8% (metals basis), foil, annealed, Thickness 0.3 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

040668-FY 040668-HQ 040668-GK
AA40668-FYEA 56 USD
AA40668-FY AA40668-HQ AA40668-GK
Niobium ≥99.8% (metals basis), foil, annealed, Thickness 0.3 mm (0.01 in)
Niobium

MDL: MFCD00011126


Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈12.9g/50x100mm Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR